Assay Design



Gene Transcripts

Gene Symbol
PPP1R42
Entrez Gene ID
704684
Gene Name
protein phosphatase 1 regulatory subunit 42
Gene Aliases
LRRC67
Chromosome Location
Chr.8: 67610197 - 67676826 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015145492.2 XP_015000978.1 - 812 75
XM_028852711.1 XP_028708544.1 - 812 75
XM_028852712.1 XP_028708545.1 - 721 75

View Full Product Details