Assay Design



Gene Transcripts

Gene Symbol
GHSR
Entrez Gene ID
694587
Gene Name
growth hormone secretagogue receptor
Gene Aliases
-
Chromosome Location
Chr.2: 77354538 - 77361766 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001084886.4 XP_001084886.1 - 829 63

View Full Product Details