Assay Design



Gene Transcripts

Gene Symbol
MICU3
Entrez Gene ID
701957
Gene Name
mitochondrial calcium uptake family member 3
Gene Aliases
EFHA2
Chromosome Location
Chr.8: 17144079 - 17236996 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028852366.1 XP_028708199.1 - 603 74
XM_028852367.1 XP_028708200.1 - 603 74
XM_028852368.1 XP_028708201.1 - 603 74
XM_028852369.1 XP_028708202.1 - 603 74
GenBank mRNA JU326223.1 - - 336 74
JV614103.1 - - 510 74

View Full Product Details