Assay Design



Gene Transcripts

Gene Symbol
SLC22A8
Entrez Gene ID
718363
Gene Name
solute carrier family 22 member 8
Gene Aliases
EGK_06087
Chromosome Location
Chr.14: 11181131 - 11204510 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194693.2 NP_001181622.1 - 1330 117
XM_001115852.4 XP_001115852.1 - 1360 117

View Full Product Details