Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_015115825.2 | XP_014971311.2 | - | 5191 | 98 |
XM_015115826.2 | XP_014971312.2 | - | 5182 | 98 | |
XM_028834035.1 | XP_028689868.1 | - | 5279 | 98 | |
XM_028834036.1 | XP_028689869.1 | - | 2675 | 98 | |
GenBank mRNA | JU343756.1 | - | - | 4746 | 98 |
JU486874.1 | - | - | 4758 | 98 |