Assay Design



Gene Transcripts

Gene Symbol
SAT2
Entrez Gene ID
716044
Gene Name
spermidine/spermine N1-acetyltransferase family member 2
Gene Aliases
EGK_08139
Chromosome Location
Chr.16: 7405362 - 7406971 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194477.1 NP_001181406.1 - 529 72
GenBank mRNA JU470851.1 - - 436 72
JV046077.1 - - 569 72
JV641233.1 - - 572 72

View Full Product Details