Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001266593.1 | NP_001253522.1 | - | 811 | 61 |
GenBank mRNA | JU333671.1 | - | - | 839 | 61 |
JU474924.1 | - | - | 838 | 61 | |
JV046582.1 | - | - | 811 | 61 | |
JV710772.1 | - | - | 840 | 61 |