Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257534.1 | NP_001244463.1 | - | 291 | 71 |
GenBank mRNA | JU320490.1 | - | - | 291 | 71 |
JU475366.1 | - | - | 382 | 71 | |
JU475367.1 | - | - | 382 | 71 | |
JV044444.1 | - | - | 335 | 71 |