Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001266741.2 | NP_001253670.1 | - | 848 | 62 |
XM_015128489.2 | XP_014983975.1 | - | 846 | 62 | |
XM_015128493.2 | XP_014983979.1 | - | 432 | 62 | |
GenBank mRNA | JU322362.1 | - | - | 855 | 62 |
JU322363.1 | - | - | 758 | 62 | |
JU471644.1 | - | - | 772 | 62 | |
JV044259.1 | - | - | 853 | 62 |