Assay Design



Gene Transcripts

Gene Symbol
CSNK1E
Entrez Gene ID
700565
Gene Name
casein kinase 1, epsilon
Gene Aliases
EGK_03040
Chromosome Location
Chr.10: 13188079 - 13241675 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261652.1 NP_001248581.1 - 1279 72
XM_015150315.2 XP_015005801.2 - 1586 72
XM_015150316.2 XP_015005802.2 - 1607 72
XM_028827234.1 XP_028683067.1 - 1586 72
XM_028827235.1 XP_028683068.1 - 1348 72
XM_028827236.1 XP_028683069.1 - 1573 72
XM_028827237.1 XP_028683070.1 - 1525 72
XM_028827238.1 XP_028683071.1 - 1287 72
GenBank mRNA JU333053.1 - - 1262 72
JU474392.1 - - 1220 72
JV046248.1 - - 1279 72
JV495557.1 - - 1265 72
JV673726.1 - - 1280 72

View Full Product Details