Assay Design



Gene Transcripts

Gene Symbol
TBC1D24
Entrez Gene ID
697106
Gene Name
TBC1 domain family member 24
Gene Aliases
-
Chromosome Location
Chr.20: 2530828 - 2535782 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257565.1 NP_001244494.1 - 1319 102
GenBank mRNA JU335621.1 - - 1319 102
JU476535.1 - - 1338 102
JV527811.1 - - 1349 102

View Full Product Details