Assay Design



Gene Transcripts

Gene Symbol
ZKSCAN5
Entrez Gene ID
718769
Gene Name
zinc finger with KRAB and SCAN domains 5
Gene Aliases
-
Chromosome Location
Chr.3: 41616135 - 41646760 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015134387.2 XP_014989873.2 - 3238 112
XM_015134389.2 XP_014989875.2 - 2046 112
XM_015134391.2 XP_014989877.2 - 3233 112
XM_015134392.2 XP_014989878.2 - 3019 112
XM_015134393.2 XP_014989879.2 - 3719 112
XM_028845558.1 XP_028701391.1 - 1907 112

View Full Product Details