Assay Design



Gene Transcripts

Gene Symbol
KDM7A
Entrez Gene ID
711830
Gene Name
lysine demethylase 7A
Gene Aliases
JHDM1D
Chromosome Location
Chr.3: 166506495 - 166601008 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015135046.2 XP_014990532.1 - 2257 86
GenBank mRNA JU326136.1 - - 2146 86
JV724219.1 - - 2058 86

View Full Product Details