Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_015151579.2 | XP_015007065.1 | - | 1080 | 84 |
XM_015151580.2 | XP_015007066.1 | - | 1212 | 84 | |
XM_015151584.2 | XP_015007070.1 | - | 1276 | 84 | |
XM_015151585.1 | XP_015007071.1 | - | 1254 | 84 | |
GenBank mRNA | JU322110.1 | - | - | 1025 | 84 |
JU470535.1 | - | - | 1020 | 84 | |
JV044367.1 | - | - | 1026 | 84 |