Assay Design



Gene Transcripts

Gene Symbol
LRIT3
Entrez Gene ID
698360
Gene Name
leucine rich repeat, Ig-like and transmembrane domains 3
Gene Aliases
-
Chromosome Location
Chr.5: 108197430 - 108223161 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001088511.4 XP_001088511.3 - 1283 72

View Full Product Details