Assay Design
Gene Transcripts
- Gene Symbol
- MAMU-DRB1
- Entrez Gene ID
-
692100
- Gene Name
- MHC class II antigen
- Gene Aliases
-
DRB1, HLA-DRB1, MAMU-DRB, Mane-DRB
- Chromosome Location
-
Chr.4: 136905344 - 136950688 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AJ601357.1 |
- |
- |
655 |
109 |
View Full Product Details