Assay Design



Gene Transcripts

Gene Symbol
CFL2
Entrez Gene ID
693875
Gene Name
cofilin 2
Gene Aliases
EGK_18105, cofilin-2
Chromosome Location
Chr.7: 96791204 - 96795570 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261184.1 NP_001248113.1 - 447 160
XM_015143527.2 XP_014999013.1 - 581 160
XM_015143528.2 XP_014999014.1 - 452 160
GenBank mRNA JU332587.1 - - 467 160
JU332588.1 - - 447 160

View Full Product Details