Assay Design



Gene Transcripts

Gene Symbol
CSNK2A1
Entrez Gene ID
714841
Gene Name
casein kinase 2 alpha 1
Gene Aliases
-
Chromosome Location
Chr.10: 66018610 - 66082026 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260877.1 NP_001247806.1 - 564 148
XM_015149293.2 XP_015004779.1 - 708 148
XM_015149294.2 XP_015004780.1 - 498 148
XM_015149295.2 XP_015004781.1 - 427 148
GenBank mRNA JU333058.1 - - 683 148
JU333059.1 - - 566 148
JU474397.1 - - 594 148
JV046251.1 - - 564 148
JV606622.1 - - 588 148

View Full Product Details