Assay Design



Gene Transcripts

Gene Symbol
MMP28
Entrez Gene ID
574250
Gene Name
matrix metallopeptidase 28
Gene Aliases
-
Chromosome Location
Chr.16: 32112299 - 32154957 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278395.2 NP_001265324.1 - 1513 117
NM_001278396.2 NP_001265325.1 - 1483 117
XM_015119023.2 XP_014974509.1 - 1702 117
XM_015119024.2 XP_014974510.1 - 2096 117
XM_028835451.1 XP_028691284.1 - 1444 117
GenBank mRNA AY145891.1 - - 185 117
JV723229.1 - - 134 117

View Full Product Details