Assay Design



Gene Transcripts

Gene Symbol
TLR6
Entrez Gene ID
574342
Gene Name
toll like receptor 6
Gene Aliases
-
Chromosome Location
Chr.5: 37964499 - 37994060 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130430.1 NP_001123902.1 - 2306 63
XM_015138136.2 XP_014993622.2 - 3763 63
XM_015138137.2 XP_014993623.2 - 2744 63
XM_028847930.1 XP_028703763.1 - 3959 63
XM_028847931.1 XP_028703764.1 - 3143 63
XM_028847932.1 XP_028703765.1 - 3005 63
XM_028847933.1 XP_028703766.1 - 2496 63
XM_028847934.1 XP_028703767.1 - 2687 63
XM_028847935.1 XP_028703768.1 - 2490 63
XM_028847936.1 XP_028703769.1 - 2608 63
GenBank mRNA AB445658.1 - - 2306 63
AY525616.1 - - 386 63
EU204941.1 - - 2315 63

View Full Product Details