Assay Design



Gene Transcripts

Gene Symbol
IFNGR1
Entrez Gene ID
100423877
Gene Name
interferon gamma receptor 1
Gene Aliases
EGK_15312
Chromosome Location
Chr.4: 40817417 - 40837449 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266300.2 NP_001253229.1 - 822 121
XM_015137238.2 XP_014992724.2 - 810 121
XM_028846596.1 XP_028702429.1 - 682 121
XM_028846597.1 XP_028702430.1 - 713 121
GenBank mRNA AF227551.1 - - 259 121
JU334343.1 - - 817 121
JU475441.1 - - 828 121
JV046958.1 - - 826 121

View Full Product Details