Assay Design



Gene Transcripts

Gene Symbol
SH2D1A
Entrez Gene ID
574156
Gene Name
SH2 domain containing 1A
Gene Aliases
EGK_20903, SAP
Chromosome Location
Chr.X: 120594638 - 120619034 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032837.1 NP_001028009.1 - 133 94
GenBank mRNA AF322912.1 - - 133 94
JU475598.1 - - 469 94
JU475599.1 - - 469 94

View Full Product Details