Assay Design



Gene Transcripts

Gene Symbol
SPA17
Entrez Gene ID
574157
Gene Name
sperm autoantigenic protein 17
Gene Aliases
-
Chromosome Location
Chr.14: 117452042 - 117473428 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172082.1 NP_001165553.1 - 384 90
GenBank mRNA AF334809.1 - - 290 90
JU323048.1 - - 403 90

View Full Product Details