Assay Design



Gene Transcripts

Gene Symbol
CCL11
Entrez Gene ID
574218
Gene Name
C-C motif chemokine ligand 11
Gene Aliases
Eotaxin, SCYA11
Chromosome Location
Chr.16: 30668726 - 30670609 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032874.1 NP_001028046.1 - 78 66
GenBank mRNA AF449270.1 - - 78 66
AY049019.1 - - 78 66

View Full Product Details