Assay Design



Gene Transcripts

Gene Symbol
CYP3A4
Entrez Gene ID
678670
Gene Name
cytochrome P450 3A64
Gene Aliases
CYP3A64
Chromosome Location
Chr.3: 41932677 - 41962305 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040414.1 NP_001035504.1 - 213 151
GenBank mRNA AY334551.1 - - 213 151
AY582531.1 - - 213 151

View Full Product Details