Assay Design



Gene Transcripts

Gene Symbol
PAEP
Entrez Gene ID
574318
Gene Name
progestagen-associated endometrial protein
Gene Aliases
-
Chromosome Location
Chr.15: 2702025 - 2717135 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032927.1 NP_001028099.1 - 140 88
XM_028834204.1 XP_028690037.1 - 1079 88
GenBank mRNA AY937248.1 - - 140 88

View Full Product Details