Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038662.1 | NP_001033751.1 | - | 2184 | 124 |
XM_028838330.1 | XP_028694163.1 | - | 2338 | 124 | |
XM_028838331.1 | XP_028694164.1 | - | 2338 | 124 | |
XM_028838333.1 | XP_028694166.1 | - | 2194 | 124 | |
GenBank mRNA | DQ227271.1 | - | - | 2184 | 124 |