Assay Design



Gene Transcripts

Gene Symbol
PFDN4
Entrez Gene ID
698809
Gene Name
prefoldin subunit 4
Gene Aliases
-
Chromosome Location
Chr.10: 89416199 - 89427413 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266117.2 NP_001253046.1 - 383 152
GenBank mRNA JU334982.1 - - 291 152
JU475995.1 - - 278 152
JV047365.1 - - 387 152

View Full Product Details