Assay Design



Gene Transcripts

Gene Symbol
BLCAP
Entrez Gene ID
708044
Gene Name
BLCAP apoptosis inducing factor
Gene Aliases
EGK_02432
Chromosome Location
Chr.10: 72775839 - 72786235 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA CB553146.1 - - 50 75
JU320479.1 - - 138 75
JU473483.1 - - 44 75
JV045999.1 - - 71 75

View Full Product Details