Assay Design



Gene Transcripts

Gene Symbol
XKRX
Entrez Gene ID
701721
Gene Name
XK related X-linked
Gene Aliases
EGK_20731
Chromosome Location
Chr.X: 97715575 - 97732335 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001090721.4 XP_001090721.1 - 2111 97
GenBank mRNA JU486597.1 - - 646 97

View Full Product Details