Assay Design
Gene Transcripts
- Gene Symbol
- H19
- Entrez Gene ID
-
704308
- Gene Name
- H19, imprinted maternally expressed transcript (non-protein coding)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.14: 2071605 - 2074240 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027325.3 |
|
- |
1685 |
69 |
GenBank mRNA |
CO579457.1 |
- |
- |
588 |
69 |
View Full Product Details