Assay Design



Gene Transcripts

Gene Symbol
PSMA2
Entrez Gene ID
701683
Gene Name
proteasome 20S subunit alpha 2
Gene Aliases
-
Chromosome Location
Chr.3: 72232225 - 72247296 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266881.1 NP_001253810.1 - 612 145
GenBank mRNA JU335507.1 - - 612 145
JV528251.1 - - 614 145

View Full Product Details