Assay Design



Gene Transcripts

Gene Symbol
PTAR1
Entrez Gene ID
700075
Gene Name
protein prenyltransferase alpha subunit repeat containing 1
Gene Aliases
-
Chromosome Location
Chr.15: 85124882 - 85174421 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265943.1 NP_001252872.1 - 340 106
XM_015117786.2 XP_014973272.1 - 406 106
XM_015117787.2 XP_014973273.1 - 423 106
XM_028834251.1 XP_028690084.1 - 393 106
XM_028834253.1 XP_028690086.1 - 397 106
XM_028834254.1 XP_028690087.1 - 397 106
GenBank mRNA JU334417.1 - - 340 106
JU334418.1 - - 340 106
JU475513.1 - - 324 106
JU475514.1 - - 324 106

View Full Product Details