Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_001085459.4 | XP_001085459.3 | - | 1519 | 78 |
XM_015123477.2 | XP_014978963.2 | - | 1082 | 78 | |
XM_015123478.2 | XP_014978964.2 | - | 1172 | 78 | |
XM_015123480.2 | XP_014978966.2 | - | 1051 | 78 | |
XM_015123483.2 | XP_014978969.2 | - | 1482 | 78 | |
XM_028839928.1 | XP_028695761.1 | - | 1355 | 78 | |
GenBank mRNA | JV664417.1 | - | - | 1191 | 78 |
JV699930.1 | - | - | 1169 | 78 |