Assay Design



Gene Transcripts

Gene Symbol
PAGE4
Entrez Gene ID
693847
Gene Name
PAGE family member 4
Gene Aliases
-
Chromosome Location
Chr.X: 49542814 - 49622367 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001082556.4 XP_001082556.1 - 404 116
XM_002806242.3 XP_002806288.1 - 356 116
XM_028842087.1 XP_028697920.1 - 419 116

View Full Product Details