Assay Design



Gene Transcripts

Gene Symbol
DNAJC27
Entrez Gene ID
694037
Gene Name
DnaJ heat shock protein family (Hsp40) member C27
Gene Aliases
EGK_05118
Chromosome Location
Chr.13: 84012212 - 84038769 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257595.1 NP_001244524.1 - 577 84
GenBank mRNA JU321934.1 - - 577 84
JU473215.1 - - 581 84
JV043655.1 - - 576 84
JV043656.1 - - 576 84

View Full Product Details