Assay Design



Gene Transcripts

Gene Symbol
HEBP1
Entrez Gene ID
697821
Gene Name
heme binding protein 1
Gene Aliases
EGK_03406, p22HBP
Chromosome Location
Chr.11: 14367267 - 14394230 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266629.1 NP_001253558.1 - 501 63
GenBank mRNA JU334749.1 - - 531 63
JV047222.1 - - 501 63
JV453164.1 - - 422 63

View Full Product Details