Assay Design



Gene Transcripts

Gene Symbol
CNIH1
Entrez Gene ID
695002
Gene Name
cornichon family AMPA receptor auxiliary protein 1
Gene Aliases
CNIH
Chromosome Location
Chr.7: 116522671 - 116538131 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193769.1 NP_001180698.1 - 381 71
XM_015143701.2 XP_014999187.1 - 445 71
GenBank mRNA JU320593.1 - - 361 71
JU472064.1 - - 332 71
JV045859.1 - - 366 71
JV528696.1 - - 345 71

View Full Product Details