Assay Design



Gene Transcripts

Gene Symbol
NMRK1
Entrez Gene ID
703868
Gene Name
nicotinamide riboside kinase 1
Gene Aliases
C15H9orf95
Chromosome Location
Chr.15: 90493972 - 90522406 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261283.1 NP_001248212.1 - 212 121
XM_015117794.2 XP_014973280.1 - 231 121
GenBank mRNA JU323108.1 - - 212 121
JU323213.1 - - 212 121

View Full Product Details