Assay Design



Gene Transcripts

Gene Symbol
CASP4
Entrez Gene ID
704864
Gene Name
caspase 4
Gene Aliases
-
Chromosome Location
Chr.14: 98189964 - 98215947 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260956.1 NP_001247885.1 - 820 74
XM_028832625.1 XP_028688458.1 - 859 74
GenBank mRNA JU473387.1 - - 820 74

View Full Product Details