Assay Design



Gene Transcripts

Gene Symbol
TNIP3
Entrez Gene ID
709178
Gene Name
TNFAIP3 interacting protein 3
Gene Aliases
-
Chromosome Location
Chr.5: 118978651 - 119058205 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015139152.2 XP_014994638.2 - 889 123

View Full Product Details