Assay Design



Gene Transcripts

Gene Symbol
TNS2
Entrez Gene ID
700813
Gene Name
tensin 2
Gene Aliases
TENC1
Chromosome Location
Chr.11: 52608417 - 52626093 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015151823.2 XP_015007309.2 - 4718 88
XM_015151825.2 XP_015007311.2 - 4448 88
XM_015151826.2 XP_015007312.2 - 4677 88
XM_028829509.1 XP_028685342.1 - 3833 88
XM_028829510.1 XP_028685343.1 - 4475 88
GenBank mRNA JU333663.1 - - 3939 88
JV646772.1 - - 3777 88

View Full Product Details