Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257621.1 | NP_001244550.1 | - | 310 | 73 |
XM_015110193.2 | XP_014965679.1 | - | 328 | 73 | |
XM_015110199.2 | XP_014965685.1 | - | 331 | 73 | |
GenBank mRNA | JU336075.1 | - | - | 310 | 73 |
JV694394.1 | - | - | 311 | 73 | |
JV721899.1 | - | - | 133 | 73 |