Assay Design



Gene Transcripts

Gene Symbol
CSNK2A1
Entrez Gene ID
714841
Gene Name
casein kinase 2 alpha 1
Gene Aliases
-
Chromosome Location
Chr.10: 66018610 - 66082026 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260877.1 NP_001247806.1 - 304 82
XM_015149293.2 XP_015004779.1 - 448 82
GenBank mRNA JU333058.1 - - 423 82
JU333059.1 - - 306 82
JU474397.1 - - 334 82
JV046251.1 - - 304 82
JV606622.1 - - 328 82

View Full Product Details