Assay Design



Gene Transcripts

Gene Symbol
AIM2
Entrez Gene ID
719260
Gene Name
absent in melanoma 2
Gene Aliases
-
Chromosome Location
Chr.1: 91777410 - 91902069 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001117183.4 XP_001117183.3 - 1101 93
XM_015112746.2 XP_014968232.2 - 1121 93
XM_028827216.1 XP_028683049.1 - 1170 93
XM_028827222.1 XP_028683055.1 - 819 93
XM_028827225.1 XP_028683058.1 - 863 93
GenBank mRNA JU325040.1 - - 779 93
JU471931.1 - - 1030 93

View Full Product Details