Assay Design



Gene Transcripts

Gene Symbol
RING1
Entrez Gene ID
718137
Gene Name
ring finger protein 1
Gene Aliases
-
Chromosome Location
Chr.4: 136293859 - 136298755 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114959.1 NP_001108431.1 - 154 63
XM_015136067.2 XP_014991553.1 - 1111 63
GenBank mRNA JU490996.1 - - 448 63
JV667246.1 - - 437 63

View Full Product Details