Assay Design



Gene Transcripts

Gene Symbol
CMTR1
Entrez Gene ID
719402
Gene Name
cap methyltransferase 1
Gene Aliases
EGK_14856, FTSJD2
Chromosome Location
Chr.4: 131935758 - 131984661 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257787.1 NP_001244716.1 - 2084 74
GenBank mRNA JU320203.1 - - 2084 74
JU471441.1 - - 2028 74
JV043937.1 - - 2150 74

View Full Product Details