Assay Design



Gene Transcripts

Gene Symbol
ARMCX3
Entrez Gene ID
704035
Gene Name
armadillo repeat containing X-linked 3
Gene Aliases
EGK_20750
Chromosome Location
Chr.X: 98419174 - 98424209 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015127819.2 XP_014983305.1 - 479 100
GenBank mRNA JU321866.1 - - 146 100
JV043803.1 - - 135 100

View Full Product Details