Assay Design



Gene Transcripts

Gene Symbol
KLHL11
Entrez Gene ID
708393
Gene Name
kelch like family member 11
Gene Aliases
-
Chromosome Location
Chr.16: 52964057 - 52982207 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001107943.4 XP_001107943.1 - 676 109
GenBank mRNA JU328201.1 - - 72 109
JV717992.1 - - 285 109

View Full Product Details