Assay Design



Gene Transcripts

Gene Symbol
BLOC1S4
Entrez Gene ID
710322
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 4
Gene Aliases
CNO
Chromosome Location
Chr.5: 6253296 - 6254582 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001091640.4 XP_001091640.2 - 622 112
GenBank mRNA JU476276.1 - - 522 112
JV636258.1 - - 570 112
JV718213.1 - - 273 112

View Full Product Details